accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 196-42-9

IUPAC Name: hexacyclo[14.6.2.0²,¹¹.0⁴,⁹.0¹³,²³.0²⁰,²⁴]tetracosa-1(22),2,4,6,8,10,12,14,16,18,20,23-dodecaene
Synonyms: hexacyclo[14.6.2.0²,¹¹.0⁴,⁹.0¹³,²³.0²⁰,²⁴]tetracosa-1(22),2,4,6,8,10,12,14,16,18,20,23-dodecaene
Molecular Weight (g/mol): 302.38
Molecular Formula: C24H14
InChi Key: AXSJLZJXXUBRBS-UHFFFAOYSA-N
SMILES: C1=CC=C2C=C3C(=CC2=C1)C=C1C=CC2=CC=CC4=CC=C3C1=C24
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results