accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFNHONHFFF

CAS RN 1960-88-9

IUPAC Name: 1,3-bis[4-(trifluoromethyl)phenyl]urea
Synonyms: 1,3-bis[4-(trifluoromethyl)phenyl]urea
Molecular Weight (g/mol): 348.25
Molecular Formula: C15H10F6N2O
InChi Key: PEXQIAHZLFTVJU-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(NC(=O)NC2=CC=C(C=C2)C(F)(F)F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results