accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19614-16-5

IUPAC Name: 1-bromo-2-(methylsulfanyl)benzene
Synonyms: 1-bromo-2-(methylsulfanyl)benzene
Molecular Weight (g/mol): 203.1
Molecular Formula: C7H7BrS
InChi Key: ALAQDUSTXPEHMH-UHFFFAOYSA-N
SMILES: CSC1=CC=CC=C1Br
Boiling Point 
  • (3)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)