CAS RN 196194-58-8
IUPAC Name:
4-chloro-2,6-difluorobenzoic acid
Synonyms:
4-chloro-2,6-difluorobenzoic acid
Molecular Weight (g/mol):
192.55
Molecular Formula:
C7H3ClF2O2
InChi Key:
ZCJKTGPZLLGECQ-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C(F)C=C(Cl)C=C1F
1
–
1
of
1
results
1
Promotions Available
4-Chloro-2,6-difluorobenzoic acid, 97%
4-Chloro-2,6-difluorobenzoic acid, 97%
CAS: 196194-58-8 Molecular Formula: C7H3ClF2O2 Molecular Weight (g/mol): 192.546 MDL Number: MFCD06203570 InChI Key: ZCJKTGPZLLGECQ-UHFFFAOYSA-N PubChem CID: 17750684 IUPAC Name: 4-chloro-2,6-difluorobenzoic acid SMILES: C1=C(C=C(C(=C1F)C(=O)O)F)Cl