accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1964-45-0

CH3SH3C

CAS RN 1964-45-0

IUPAC Name: 3,5-dimethyl-1-benzothiophene
Synonyms: 3,5-dimethyl-1-benzothiophene
Molecular Weight (g/mol): 162.25
Molecular Formula: C10H10S
InChi Key: UQJSHXYNNVEGMQ-UHFFFAOYSA-N
SMILES: CC1=CSC2=CC=C(C)C=C12
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results