accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19648-86-3

IUPAC Name: bis(1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one) manganese
Synonyms: bis(1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one) manganese
Molecular Weight (g/mol): 471.06
Molecular Formula: C10H4F12MnO4
InChi Key: DMABZFSNYKYMMQ-UHFFFAOYSA-N
SMILES: [Mn].OC(=CC(=O)C(F)(F)F)C(F)(F)F.OC(=CC(=O)C(F)(F)F)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results