accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19652-13-2

IUPAC Name: 3-(4-methoxyphenyl)-1-phenyl-1H-pyrazol-5-amine
Synonyms: 5-(4-methoxyphenyl)-2-phenylpyrazol-3-amine
Molecular Weight (g/mol): 265.32
Molecular Formula: C16H15N3O
InChi Key: LMLGFNHDEGGHOJ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=NN(C(N)=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results