CAS RN 19654-19-4
IUPAC Name:
2-(4-bromophenyl)-1,3-benzothiazole
Synonyms:
2-(4-bromophenyl)-1,3-benzothiazole
Molecular Weight (g/mol):
290.18
Molecular Formula:
C13H8BrNS
InChi Key:
FQIRBKKYMJKENC-UHFFFAOYSA-N
SMILES:
BrC1=CC=C(C=C1)C1=NC2=CC=CC=C2S1
1
–
1
of
1
results
1
2-(4-Bromophenyl)benzothiazole 98.0+%, TCI America™
CAS: 19654-19-4 Molecular Formula: C13H8BrNS Molecular Weight (g/mol): 290.18 MDL Number: MFCD00579199 InChI Key: FQIRBKKYMJKENC-UHFFFAOYSA-N Synonym: 1-(2-Benzothiazolyl)-4-bromobenzene PubChem CID: 2320602 IUPAC Name: 2-(4-bromophenyl)-1,3-benzothiazole SMILES: BrC1=CC=C(C=C1)C1=NC2=CC=CC=C2S1