accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NH

CAS RN 19678-58-1

IUPAC Name: 4-(propan-2-yl)piperidine
Synonyms: 4-isopropylpiperidine
Molecular Weight (g/mol): 127.23
Molecular Formula: C8H17N
InChi Key: YBPWIUSXQXYTSR-UHFFFAOYSA-N
SMILES: CC(C)C1CCNCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results