accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH2

CAS RN 1968-40-7

IUPAC Name: ethyl pent-4-enoate
Synonyms: ethyl pent-4-enoate
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: PTVSRINJXWDIKP-UHFFFAOYSA-N
SMILES: CCOC(=O)CCC=C
Percent Purity 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results