CAS RN 19689-95-3
IUPAC Name:
methylamino acetate hydrochloride
Synonyms:
methylamino acetate hydrochloride
Molecular Weight (g/mol):
125.55
Molecular Formula:
C3H8ClNO2
InChi Key:
ZTMICKVBCKWFHL-UHFFFAOYSA-N
SMILES:
Cl.CNOC(C)=O
1
–
1
of
1
results
1
O-Acetyl-N-methylhydroxylamine Hydrochloride 98.0+%, TCI America™
CAS: 19689-95-3 Molecular Formula: C3H8ClNO2 Molecular Weight (g/mol): 125.552 InChI Key: ZTMICKVBCKWFHL-UHFFFAOYSA-N PubChem CID: 13675437 IUPAC Name: methylamino acetate;hydrochloride SMILES: CC(=O)ONC.Cl