accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19690-59-6

IUPAC Name: hydrogen (4-chloro-2-methylphenyl)hydrazine chloride
Synonyms: hydrogen (4-chloro-2-methylphenyl)hydrazine chloride
Molecular Weight (g/mol): 193.07
Molecular Formula: C7H10Cl2N2
InChi Key: VBVZXWFQHFXJJK-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=CC(Cl)=CC=C1NN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results