accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHO

CAS RN 19694-02-1

IUPAC Name: pyrene-1-carboxylic acid
Synonyms: pyrene-1-carboxylic acid
Molecular Weight (g/mol): 246.27
Molecular Formula: C17H10O2
InChi Key: HYISVWRHTUCNCS-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)