accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19740-72-8

IUPAC Name: N'-[(propan-2-yloxy)carbonyl](propan-2-yloxy)carbohydrazide
Synonyms: N'-(isopropoxycarbonyl)isopropoxycarbohydrazide
Molecular Weight (g/mol): 204.23
Molecular Formula: C8H16N2O4
InChi Key: FBZULTVJWVCJQV-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)NNC(=O)OC(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results