accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONHS(R)HNOOCH3CH3H3CHOO

CAS RN 19746-37-3

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-[(acetamidomethyl)sulfanyl]propanoic acid
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]-3-[(acetamidomethyl)sulfanyl]propanoic acid
Molecular Weight (g/mol): 292.35
Molecular Formula: C11H20N2O5S
InChi Key: HLCTYBOTPCIHTG-QMMMGPOBSA-N
SMILES: CC(=O)NCSC[C@H](NC(=O)OC(C)(C)C)C(O)=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)