accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19752-55-7

IUPAC Name: 1-bromo-3,5-dichlorobenzene
Synonyms: benzene, 1-bromo-3,5-dichloro-
Molecular Weight (g/mol): 225.89
Molecular Formula: C6H3BrCl2
InChi Key: DZHFFMWJXJBBRG-UHFFFAOYSA-N
SMILES: ClC1=CC(Br)=CC(Cl)=C1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (1)
  • (3)
  • (3)
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
Boiling Point 
  • (5)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (5)
  • (4)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (5)
  • (1)
  • (3)