accessibility menu, dialog, popup

UserName

Viewing profile as user:
ZrNCH3H3CNCH3CH3NCH3H3CNCH3H3C

CAS RN 19756-04-8

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 267.53
Molecular Formula: C8H24N4Zr
InChi Key: DWCMDRNGBIZOQL-UHFFFAOYSA-N
SMILES: [Zr+4].C[N-]C.C[N-]C.C[N-]C.C[N-]C
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)