accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONHOHONCH3OO

CAS RN 197632-76-1

IUPAC Name: 6-{[(tert-butoxy)carbonyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)hexanoic acid
Synonyms: 6-[(tert-butoxycarbonyl)amino]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl](methyl)amino}hexanoic acid
Molecular Weight (g/mol): 482.58
Molecular Formula: C27H34N2O6
InChi Key: JMBKBGOKNZZJQA-UHFFFAOYNA-N
SMILES: CN(C(CCCCNC(=O)OC(C)(C)C)C(O)=O)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results