CAS RN 197770-48-2
IUPAC Name:
4-[2-(4-fluorophenyl)ethynyl]phenol
Synonyms:
4-[2-(4-fluorophenyl)ethynyl]phenol
Molecular Weight (g/mol):
212.22
Molecular Formula:
C14H9FO
InChi Key:
LIPHCUGEWJPCQB-UHFFFAOYSA-N
SMILES:
OC1=CC=C(C=C1)C#CC1=CC=C(F)C=C1
1
–
1
of
1
results
1
4-(4-Fluorophenylethynyl)phenol 98.0+%, TCI America™
CAS: 197770-48-2 Molecular Formula: C14H9FO Molecular Weight (g/mol): 212.223 MDL Number: MFCD02093472 InChI Key: LIPHCUGEWJPCQB-UHFFFAOYSA-N Synonym: 1-(4-Fluorophenyl)-2-(4-hydroxyphenyl)acetylene PubChem CID: 2782365 IUPAC Name: 4-[2-(4-fluorophenyl)ethynyl]phenol SMILES: C1=CC(=CC=C1C#CC2=CC=C(C=C2)F)O