accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1978-38-7

H3CNHF

CAS RN 1978-38-7

IUPAC Name: 2-fluoro-N-methylaniline
Synonyms: 2-fluoro-N-methylaniline
Molecular Weight (g/mol): 125.15
Molecular Formula: C7H8FN
InChi Key: LDVAIJZDACHGML-UHFFFAOYSA-N
SMILES: CNC1=CC=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results