accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19780-11-1

IUPAC Name: 3-[(2E)-dodec-2-en-1-yl]oxolane-2,5-dione
Synonyms: 3-[(2E)-dodec-2-en-1-yl]oxolane-2,5-dione
Molecular Weight (g/mol): 266.38
Molecular Formula: C16H26O3
InChi Key: UYCICMIUKYEYEU-ZHACJKMWNA-N
SMILES: CCCCCCCCC\C=C\CC1CC(=O)OC1=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)