CAS RN 19780-12-2
IUPAC Name:
dodec-5-yne
Synonyms:
dodec-5-yne
Molecular Weight (g/mol):
166.31
Molecular Formula:
C12H22
InChi Key:
JQPXWRQCSIYOOP-UHFFFAOYSA-N
SMILES:
CCCCCCC#CCCCC
1
–
1
of
1
results
1
5-Dodecyne 98.0+%, TCI America™
CAS: 19780-12-2 Molecular Formula: C12H22 Molecular Weight (g/mol): 166.31 MDL Number: MFCD00041661 InChI Key: JQPXWRQCSIYOOP-UHFFFAOYSA-N PubChem CID: 140583 IUPAC Name: dodec-5-yne SMILES: CCCCCCC#CCCCC