accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO

CAS RN 19780-16-6

IUPAC Name: 2-octadecyloxirane
Synonyms: 2-octadecyloxirane
Molecular Weight (g/mol): 296.54
Molecular Formula: C20H40O
InChi Key: BHZBVWCLMYQFQX-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCCCCCC1CO1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results