accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19780-79-1

IUPAC Name: 2-hexyloctan-1-ol
Synonyms: 2-hexyloctan-1-ol
Molecular Weight (g/mol): 214.39
Molecular Formula: C14H30O
InChi Key: QNMCWJOEQBZQHB-UHFFFAOYSA-N
SMILES: CCCCCCC(CO)CCCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results