accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19788-37-5

IUPAC Name: 4-(chloromethyl)-3,5-dimethyl-1,2-oxazole
Synonyms: 4-(chloromethyl)-3,5-dimethyl-1,2-oxazole
Molecular Weight (g/mol): 145.59
Molecular Formula: C6H8ClNO
InChi Key: NIFAUKBQIAURIM-UHFFFAOYSA-N
SMILES: CC1=C(CCl)C(C)=NO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results