accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOBOHN

CAS RN 197958-29-5

IUPAC Name: (pyridin-2-yl)boronic acid
Synonyms: pyridin-2-ylboronic acid
Molecular Weight (g/mol): 122.92
Molecular Formula: C5H6BNO2
InChi Key: UMLDUMMLRZFROX-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results