accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NHCl

CAS RN 19798-77-7

IUPAC Name: 4-amino-3-chloropyridin-1-ium
Synonyms: 4-amino-3-chloropyridin-1-ium
Molecular Weight (g/mol): 129.57
Molecular Formula: C5H6ClN2
InChi Key: KJBKPRMEMJKXDV-UHFFFAOYSA-O
SMILES: NC1=CC=[NH+]C=C1Cl
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)