accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2BrHN

CAS RN 19798-81-3

IUPAC Name: 2-amino-6-bromopyridin-1-ium
Synonyms: 2-amino-6-bromopyridin-1-ium
Molecular Weight (g/mol): 174.02
Molecular Formula: C5H6BrN2
InChi Key: BKLJUYPLUWUEOQ-UHFFFAOYSA-O
SMILES: NC1=CC=CC(Br)=[NH+]1
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (3)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)