accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONN

CAS RN 198084-12-7

IUPAC Name: 4-(pyrimidin-5-yl)benzaldehyde
Synonyms: 4-(pyrimidin-5-yl)benzaldehyde
Molecular Weight (g/mol): 184.2
Molecular Formula: C11H8N2O
InChi Key: BXNXTOLXLPQMHS-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(C=C1)C1=CN=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results