accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CO

CAS RN 19814-75-6

IUPAC Name: 9,9-dimethyl-9H-xanthene
Synonyms: 9,9-dimethylxanthene
Molecular Weight (g/mol): 210.28
Molecular Formula: C15H14O
InChi Key: MTVNAPYHLASOSX-UHFFFAOYSA-N
SMILES: CC1(C)C2=CC=CC=C2OC2=CC=CC=C12
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)