accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrIBr

CAS RN 19821-80-8

IUPAC Name: 1,3-dibromo-2-iodobenzene
Synonyms: 1,3-dibromo-2-iodobenzene
Molecular Weight (g/mol): 361.8
Molecular Formula: C6H3Br2I
InChi Key: OIFRMZVTJQAPIF-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(Br)=C1I
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results