accessibility menu, dialog, popup

UserName

Viewing profile as user:
HN

CAS RN 198275-79-5

IUPAC Name: N-phenyl-[1,1'-biphenyl]-3-amine
Synonyms: N-phenyl-[1,1'-biphenyl]-3-amine
Molecular Weight (g/mol): 245.33
Molecular Formula: C18H15N
InChi Key: QJAYWJUCAONYLG-UHFFFAOYSA-N
SMILES: N(C1=CC=CC=C1)C1=CC=CC(=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results