accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCl

CAS RN 19832-87-2

IUPAC Name: 2-chloro-5-methylcyclohexa-2,5-diene-1,4-dione
Synonyms: 2-chloro-5-methylcyclohexa-2,5-diene-1,4-dione
Molecular Weight (g/mol): 156.57
Molecular Formula: C7H5ClO2
InChi Key: BIUBEGIHROOPTQ-UHFFFAOYSA-N
SMILES: CC1=CC(=O)C(Cl)=CC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results