accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNS

CAS RN 198334-38-2

IUPAC Name: 4-(thiophen-2-yl)piperidine
Synonyms: 4-(thiophen-2-yl)piperidine
Molecular Weight (g/mol): 167.27
Molecular Formula: C9H13NS
InChi Key: SLQVNTKKWCOXKW-UHFFFAOYSA-N
SMILES: C1CC(CCN1)C1=CC=CS1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results