accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOO

CAS RN 19836-78-3

IUPAC Name: 3-methyl-1,3-oxazolidin-2-one
Synonyms: 2-oxazolidinone, 3-methyl-
Molecular Weight (g/mol): 101.11
Molecular Formula: C4H7NO2
InChi Key: VWIIJDNADIEEDB-UHFFFAOYSA-N
SMILES: CN1CCOC1=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)