accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COClCl

CAS RN 1984-59-4

IUPAC Name: 1,2-dichloro-3-methoxybenzene
Synonyms: 1,2-dichloro-3-methoxybenzene
Molecular Weight (g/mol): 177.02
Molecular Formula: C7H6Cl2O
InChi Key: HFEASCCDHUVYKU-UHFFFAOYSA-N
SMILES: COC1=CC=CC(Cl)=C1Cl
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)