accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH2OOH3CCH3OOCH3H2C

CAS RN 1985-51-9

IUPAC Name: 2,2-dimethyl-3-[(2-methylprop-2-enoyl)oxy]propyl 2-methylprop-2-enoate
Synonyms: neopentylglycol dimethacrylate neopentyl glycol dimethacrylate
Molecular Weight (g/mol): 240.3
Molecular Formula: C13H20O4
InChi Key: ULQMPOIOSDXIGC-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OCC(C)(C)COC(=O)C(C)=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results