accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH(R)OHONHOO

CAS RN 198544-94-4

IUPAC Name: (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-6-{[(4-methylphenyl)diphenylmethyl]amino}hexanoic acid
Synonyms: (2R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-6-{[(4-methylphenyl)diphenylmethyl]amino}hexanoic acid|FMOC-D-LYS(MTT)-OH
Molecular Weight (g/mol): 624.78
Molecular Formula: C41H40N2O4
InChi Key: YPTNAIDIXCOZAJ-KXQOOQHDSA-N
SMILES: CC1=CC=C(C=C1)C(NCCCC[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)