accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOONHNH(R)H3C

CAS RN 198545-20-9

IUPAC Name: (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-{[(4-methylphenyl)diphenylmethyl]amino}pentanoic acid
Synonyms: (2R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-5-{[(4-methylphenyl)diphenylmethyl]amino}pentanoic acid
Molecular Weight (g/mol): 610.75
Molecular Formula: C40H38N2O4
InChi Key: RRYSGISHZIBOJC-DIPNUNPCSA-N
SMILES: CC1=CC=C(C=C1)C(NCCC[C@@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)