accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 198560-44-0

IUPAC Name: (2S)-3-(3-chlorophenyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: (2S)-3-(3-chlorophenyl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 421.88
Molecular Formula: C24H20ClNO4
InChi Key: UOZAKKJRIKXQPY-QFIPXVFZSA-N
SMILES: OC(=O)[C@H](CC1=CC=CC(Cl)=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (1)
Quantity 
  • (3)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)