accessibility menu, dialog, popup

UserName

Viewing profile as user:
FOOHOONH(S)

CAS RN 198560-68-8

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(3-fluorophenyl)propanoic acid
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(3-fluorophenyl)propanoic acid
Molecular Weight (g/mol): 405.43
Molecular Formula: C24H20FNO4
InChi Key: DWSDVARCJDOADL-QFIPXVFZSA-N
SMILES: OC(=O)[C@H](CC1=CC(F)=CC=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results