accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 198561-86-3

ClOOOHOOONHNH(S)

CAS RN 198561-86-3

IUPAC Name: (2S)-5-({[(2-chlorophenyl)methoxy]carbonyl}amino)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoic acid
Synonyms: (2S)-5-({[(2-chlorophenyl)methoxy]carbonyl}amino)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}pentanoic acid
Molecular Weight (g/mol): 522.98
Molecular Formula: C28H27ClN2O6
InChi Key: CVAOOYINJZPDDP-VWLOTQADSA-N
SMILES: OC(=O)[C@H](CCCNC(=O)OCC1=C(Cl)C=CC=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)