accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSOCH3

CAS RN 1986-89-6

IUPAC Name: 1-(octane-1-sulfinyl)octane
Synonyms: 1-(octane-1-sulfinyl)octane
Molecular Weight (g/mol): 274.51
Molecular Formula: C16H34OS
InChi Key: VWCFQNQVNVMFGV-UHFFFAOYSA-N
SMILES: CCCCCCCCS(=O)CCCCCCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results