accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCH3CH3HS

CAS RN 19872-52-7

IUPAC Name: 4-methyl-4-sulfanylpentan-2-one
Synonyms: 4-methyl-4-sulfanylpentan-2-one
Molecular Weight (g/mol): 132.22
Molecular Formula: C6H12OS
InChi Key: QRNZMFDCKKEPSX-UHFFFAOYSA-N
SMILES: CC(=O)CC(C)(C)S
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)