accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3COO(R)NH2

CAS RN 19883-41-1

IUPAC Name: hydrogen methyl 2-(2-aminophenyl)acetate chloride
Synonyms: hydrogen methyl 2-(2-aminophenyl)acetate chloride
Molecular Weight (g/mol): 201.65
Molecular Formula: C9H12ClNO2
InChi Key: OZMUKUBEEKGADM-UHFFFAOYSA-N
SMILES: [H+].[Cl-].COC(=O)CC1=CC=CC=C1N
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)