accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3CH3CH3CH3CH3

CAS RN 1989-52-2

IUPAC Name: 9a,11a-dimethyl-1-(6-methylheptan-2-yl)-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl tetradecanoate
Synonyms: 9a,11a-dimethyl-1-(6-methylheptan-2-yl)-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9bH,10H,11H-cyclopenta[a]phenanthren-7-yl tetradecanoate
Molecular Weight (g/mol): 597.03
Molecular Formula: C41H72O2
InChi Key: SJDMTGSQPOFVLR-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCC(=O)OC1CCC2(C)C3CCC4(C)C(CCC4C3CC=C2C1)C(C)CCCC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results