accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONS

CAS RN 198904-53-9

IUPAC Name: 4-(1,3-thiazol-2-yl)benzaldehyde
Synonyms: 4-(1,3-thiazol-2-yl)benzaldehyde
Molecular Weight (g/mol): 189.23
Molecular Formula: C10H7NOS
InChi Key: LQLBILPEELCFQI-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(C=C1)C1=NC=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results