accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 198976-43-1

IUPAC Name: piperidin-3-ol hydrochloride
Synonyms: piperidin-3-ol hydrochloride
Molecular Weight (g/mol): 137.61
Molecular Formula: C5H12ClNO
InChi Key: VLECDMDGMKPUSK-UHFFFAOYNA-N
SMILES: Cl.OC1CCCNC1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results