accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 199106-97-3

IUPAC Name: N-ethyl-2-methoxy-N-(3-methylphenyl)benzamide
Synonyms: N-ethyl-2-methoxy-N-(3-methylphenyl)benzamide
Molecular Weight (g/mol): 269.34
Molecular Formula: C17H19NO2
InChi Key: FYICKJLKZRYUEZ-UHFFFAOYSA-N
SMILES: CCN(C(=O)C1=CC=CC=C1OC)C1=CC=CC(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results