accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHNCH3OOCH3CH3H3C

CAS RN 19914-38-6

IUPAC Name: 2-{[(tert-butoxy)carbonyl](methyl)amino}propanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)(methyl)amino]propanoic acid
Molecular Weight (g/mol): 203.24
Molecular Formula: C9H17NO4
InChi Key: VLHQXRIIQSTJCQ-UHFFFAOYNA-N
SMILES: CC(N(C)C(=O)OC(C)(C)C)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results